2,4-Dihydroxy-5-methoxypyrimidine ≥95%

Supplier: BeanTown Chemical

Synonyms: 5-Methoxy-2,4-pyrimidinediol

128705-1G 128705-5G
BT128705-1GEA 86 USD
BT128705-1G BT128705-5G
2,4-Dihydroxy-5-methoxypyrimidine ≥95%
2,4-Dihydroxy-5-methoxypyrimidine

CAS: 6623-81-0; EC No: 229-580-5; MDL No: MFCD03428618 Solid; Molecular Formula: C5H6N2O3; MW: 142.11


  • Solid

Formula: C₅H₆N₂O₃
MW: 142.1100 g/mol
MDL Number: MFCD00187903
CAS Number: 6623-81-0
EINECS: 229-580-5

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR