6-Chloro-4-quinazolone

Supplier: BeanTown Chemical

Synonyms: 6-Chloroquinazolin-4-ol

128700-1G 128700-5G
BT128700-1GEA 61 USD
BT128700-1G BT128700-5G
6-Chloro-4-quinazolone
6-Chloro-4-quinazolone

CAS: 16064-14-5; MDL No: MFCD01686365 Solid; Molecular Formula: C8H5ClN2O; MW: 180.59


  • Solid

Formula: C₈H₅ClN₂O
MW: 180.5900 g/mol
MDL Number: MFCD01686365
CAS Number: 16064-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR