Silica gel, Pore size: 60 Å 0.062-0.105mm; 150-230 mesh

Supplier: BeanTown Chemical

128675-1KG 128675-250G 128675-50G
BT128675-1KGEA 251 USD
BT128675-1KG BT128675-250G BT128675-50G
Silica gel, Pore size: 60 Å 0.062-0.105mm; 150-230 mesh
Silica gel

CAS: 63231-67-4; EC No: 231-545-4; MDL No: MFCD00011232 Powder, S.A. 500-600 m2/g Melting Point: 1600°; Boiling Point: 2230° Hygroscopic


  • Hygroscopic
  • Powder, S.A. 500-600 m²/g

Formula: SiO₂
Boiling Pt: 2230 °C
Melting Pt: >1600 °C
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR