Tolbutamide ≥98%

Supplier: BeanTown Chemical

Synonyms: N-(Butylcarbamoyl)-4-methylbenzenesulphonamide

128590-10G 128590-250G 128590-50G
BT128590-10GEA 54 USD
BT128590-10G BT128590-250G BT128590-50G
Tolbutamide ≥98%
Tolbutamide

CAS: 64-77-7; EC No: 200-594-3; MDL No: MFCD00027169; RTECS: YS4550000 Solid; Molecular Formula: C12H18N2O3S; MW: 270.35 Melting Point: 128-130°


  • RTECS: YS4550000
  • Solid

Formula: C₁₂H₁₈N₂O₃S
MW: 270.35
Melting Pt: 128…130 °C
MDL Number: MFCD00027169
CAS Number: 64-77-7
EINECS: 200-594-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR