3,3',5,5'-Tetra-tert-butyl-4,4'-diphenoquinone ≥98%
Supplier: BeanTown Chemical
128585-1G
128585-5G
BT128585-1GEA
57
USD
BT128585-1G
BT128585-5G
3,3',5,5'-Tetra-tert-butyl-4,4'-diphenoquinone ≥98%
3,3',5,5'-Tetra-tert-butyl-4,4'-diphenoquinone
CAS: 2455-14-3; EC No: 219-527-4; MDL No: MFCD00051798
Solid; Molecular Formula: C28H40O2; MW: 408.63
- Solid
Formula:
C₂₈H₄₀O₂ |
MDL Number:
MFCD00051798 CAS Number: 2455-14-3 EINECS: 219-527-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...