Antimony ≥99.999% (trace metals basis), spherical shots 1-5 mm

Supplier: BeanTown Chemical


Ampuled under nitrogen

128555-10G 128555-50G
BT128555-10GEA 239 USD
BT128555-10G BT128555-50G
Antimony ≥99.999% (trace metals basis), spherical shots 1-5 mm
Antimony

CAS: 7440-36-0; EC No: 231-146-5; MDL No: MFCD00134030; RTECS: CC4025000 Solid; Molecular Formula: Sb; MW: 121.76 Melting Point: 630°; Boiling Point: 1635° Density (g/mL): 6.69

Formula: Sb
MW: 121.76 g/mol
Boiling Pt: 1635 °C
Melting Pt: 630 °C
Density: 6.69 g/mL
MDL Number: MFCD00134030
CAS Number: 7440-36-0
EINECS: 231-146-5
UN: 2871
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99.999%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR