(±)-Cbz-α-phosphonoglycine trimethyl ester ≥97%

Supplier: BeanTown Chemical

Synonyms: N-Benzyloxycarbonyl-2-phosphonoglycine trimethyl ester, (±)-Benzyloxycarbonyl-α-phosphonoglycine trimethyl ester, (±)-Methyl 2-benzyloxycarbonylamino-2-(dimethoxyphosphinyl)acetate, Methyl 2-{[(benzyloxy)carbonyl]amino}-2-(dimethoxyphosphoryl)acetate, Methyl 2-(((benzyloxy)carbonyl)amino)-2-(dimethoxyphosphoryl)acetate

128540-1G 128540-25G 128540-5G
BT128540-1GEA 62 USD
BT128540-1G BT128540-25G BT128540-5G
(±)-Cbz-α-phosphonoglycine trimethyl ester ≥97%
(±)-Cbz-α-phosphonoglycine trimethyl ester

CAS: 88568-95-0; MDL No: MFCD00043304 Powder; Molecular Formula: C13H18NO7P; MW: 331.26 Melting Point: 76-80°


  • Powder

Formula: C₁₃H₁₈NO₇P
MW: 331.27 g/mol
Melting Pt: 76…80 °C
MDL Number: MFCD00043304
CAS Number: 88568-95-0

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR