Germanium(IV) oxide ≥99.999% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Germanium dioxide
128535-100G
128535-25G
128535-5G
BT128535-100GEA
523
USD
BT128535-100G
BT128535-25G
BT128535-5G
Germanium(IV) oxide ≥99.999% (trace metals basis)
Germanium(IV) oxide
CAS: 1310-53-8; EC No: 215-180-8; MDL No: MFCD00011030
Powder; Molecular Formula: GeO2; MW: 104.64
Melting Point:
- Powder
Formula:
GeO₂ MW: 104.61 g/mol Melting Pt: >400 °C Density: 3.637 g/mL |
MDL Number:
MFCD00011030 CAS Number: 1310-53-8 EINECS: 215-180-8 |
Specification Test Results
Assay | 99.999% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...