Terbium(III) chloride, anhydrous ≥99.99%
Supplier: BeanTown Chemical
Ampuled under argon
128525-10G
128525-1G
BT128525-10GEA
1049
USD
BT128525-10G
BT128525-1G
Terbium(III) chloride, anhydrous ≥99.99%
Terbium(III) chloride
CAS: 10042-88-3; EC No: 233-132-4; MDL No: MFCD00011258
Powder; Linear Formula: TbCl3; MW: 265.29
Melting Point: 588°
Density (g/mL): 4.35
Hygroscopic
- Hygroscopic
- Powder
Formula:
Cl₃Tb MW: 265.28 Melting Pt: 588 Density: 4.35 g/mL |
MDL Number:
MFCD00011258 CAS Number: 10042-88-3 EINECS: 233-132-4 Merck Index: 13,09087 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...