DL-Isoleucine ≥99%

Supplier: BeanTown Chemical

128515-100G 128515-25G
BT128515-100GEA 180.62 USD
BT128515-100G BT128515-25G
DL-Isoleucine ≥99%
DL-Isoleucine

CAS: 443-79-8; EC No: 207-139-8; MDL No: MFCD00004268 Powder; Molecular Formula: C6H13NO2; MW: 131.18 Melting Point: 290° (decomposes)


  • Powder

Formula: C₆H₁₃NO₂
Melting Pt: 290 °C (dec.)
MDL Number: MFCD00004268
CAS Number: 443-79-8
EINECS: 207-139-8

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR