(S)-(+)-1-(2-Chlorophenyl)-1,2-ethanediol ≥96%
Supplier: BeanTown Chemical
Synonyms:
(1S)-1-(2-chlorophenyl)ethane-1,2-diol
128510-1G
128510-250MG
128510-5G
BT128510-1GEA
158.63
USD
BT128510-1G
BT128510-250MG
BT128510-5G
(S)-(+)-1-(2-Chlorophenyl)-1,2-ethanediol ≥96%
(S)-(+)-1-(2-Chlorophenyl)-1,2-ethanediol
CAS: 133082-13-0; MDL No: MFCD01863241
Solid; Molecular Formula: C8H9ClO2 ; MW: 172.61
Melting Point: 68-75°; Boiling Point: 293°; Flash point: 108°C (226°F)
Optical Rotation: [α]20/D +70°, c = 0.2 in chloroform
- Optical rotation: [α]20/D +70°, c = 0.2 in chloroform
- Solid
Formula:
C₈H₉ClO₂ MW: 172.608 g/mol Boiling Pt: 293 °C Melting Pt: 68…75 °C Flash Pt: 108 °C (226 °F) |
MDL Number:
MFCD01863241 CAS Number: 133082-13-0 |
Specification Test Results
Assay | 96% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...