2-Hydroxyquinoline-4-carboxylic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-Hydroxycinchoninic Acid

128485-1G 128485-5G
BT128485-1GEA 58 USD
BT128485-1G BT128485-5G
2-Hydroxyquinoline-4-carboxylic acid ≥98%
2-Hydroxyquinoline-4-carboxylic acid

CAS: 15733-89-8; EC No: 239-827-9; MDL No: MFCD00464512; RTECS: GD3869000 Powder; Molecular Formula: C10H7NO3; MW: 189.17


  • RTECS: GD3869000
  • Powder

Formula: C₁₀H₇NO₃
MW: 189.17 g/mol
MDL Number: MFCD00023942
CAS Number: 15733-89-8
EINECS: 239-827-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR