Manganese(III) acetate hydrate ≥96%

Supplier: BeanTown Chemical
Warning

128465-10G 128465-50G
BT128465-10GEA 87.57 USD
BT128465-10G BT128465-50G
Manganese(III) acetate hydrate ≥96%
Manganese(III) acetate hydrate

CAS: 19513-05-4; EC No: 213-602-5; MDL No: MFCD00150022 Crystalline/Powder; Linear Formula: (CH3COO)3Mn·xH2O; MW: 232.07 (anhydrous) Density (g/mL): 1.59 Moisture Sensitive


  • Crystalline/Powder
  • Moisture Sensitive

Formula: C₆H₉MnO₆·xH₂O
Density: 1.59 g/mL
MDL Number: MFCD00150022
CAS Number: 19513-05-4
EINECS: 213-602-5

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR