Divinylbenzene (mixture of isomers) 55% stabilized, Technical Grade
Supplier: BeanTown Chemical
Synonyms:
DVB, Divinylbenzene
128460-1KG
128460-250G
128460-50G
BT128460-1KGEA
169
USD
BT128460-1KG
BT128460-250G
BT128460-50G
Divinylbenzene (mixture of isomers) 55% stabilized, Technical Grade
Divinylbenzene (mixture of isomers)
CAS: 1321-74-0; EC No: 215-325-5; MDL No: MFCD01778656; RTECS: CZ9370000
Liquid; Linear Formula: C6H4(CH=CH2)2; Molecular Formula: C10H10; MW: 130.19
Boiling Point: 195°; Flash point: 62°C (144°F)
Density (g/mL): 0.912; Refractive Index: 1.561
- RTECS: CZ9370000
- Refractive index: 1.561
- Liquid
Formula:
C₁₀H₁₀ MW: 130.19 g/mol Boiling Pt: 195 °C Density: 0.912 g/mL Flash Pt: 62 °C (144 °F) |
MDL Number:
MFCD01778656 CAS Number: 1321-74-0 EINECS: 215-325-5 |
Specification Test Results
Assay | 55% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...