3-Bromo-1-propanol ≥97%

Supplier: BeanTown Chemical

Synonyms: 3-Bromopropan-1-ol, 1-Bromo-3-hydroxypropane

128440-100G 128440-25G 128440-5G
BT128440-100GEA 292.35 USD
BT128440-100G BT128440-25G BT128440-5G
3-Bromo-1-propanol ≥97%
3-Bromo-1-propanol

CAS: 627-18-9; EC No: 210-986-6; MDL No: MFCD00002942; RTECS: UA7385000 Liquid; Linear Formula: Br(CH2)3OH; MW: 138.99 Boiling Point: 62°/5 mmHg; Flash point: 101°C (214°F) Density (g/mL): 1.537; Refractive Index: 1.488


  • RTECS: UA7385000
  • Refractive index: 1.488
  • Liquid

Formula: BrH₂CCH₂CH₂OH
MW: 138.99 g/mol
Boiling Pt: 62 °C/5 mmHg
Density: 1.537 g/mL
Flash Pt: 101 °C (214 °F)
MDL Number: MFCD00002942
CAS Number: 627-18-9
EINECS: 210-986-6

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR