5-Bromoquinoline ≥97%
Supplier: BeanTown Chemical
128435-1G
128435-250MG
BT128435-1GEA
175
USD
BT128435-1G
BT128435-250MG
5-Bromoquinoline ≥97%
5-Bromoquinoline
CAS: 4964-71-0; MDL No: MFCD00234481
Solid; Molecular Formula: C9H6BrN ; MW: 208.05
Melting Point: 43-48°
- Solid
Formula:
C₉H₆BrN MW: 208.06 g/mol Melting Pt: 43…48 °C |
MDL Number:
MFCD00234481 CAS Number: 4964-71-0 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...