Tellurium dioxide ≥99.9995%
Supplier: BeanTown Chemical
128420-100G
128420-25G
128420-500G
BT128420-100GEA
419
USD
BT128420-100G
BT128420-25G
BT128420-500G
Tellurium dioxide ≥99.9995%
Tellurium dioxide
CAS: 7446-07-3; EC No: 231-193-1; MDL No: MFCD00011263; RTECS: WY2675000
Powder; Linear Formula: TeO2; MW: 159.60
Melting Point: 733°
Density (g/mL): 5.67
- RTECS: WY2675000
- Powder
Formula:
O₂Te MW: 159.6 g/mol Boiling Pt: 1245 Melting Pt: 773 Density: 5.67 g/mL |
MDL Number:
MFCD00011263 CAS Number: 7446-07-3 EINECS: 231-193-1 Merck Index: 14,09123 |
Specification Test Results
Assay | 99.9995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...