Tripotassium orthophosphate, anhydrous ≥97%

Supplier: BeanTown Chemical

Synonyms: Potassium phosphate tribasic, Potassium phosphate

128380-1KG 128380-250G 128380-5KG
BT128380-1KGEA 141.43 USD
BT128380-1KG BT128380-250G BT128380-5KG
Tripotassium orthophosphate, anhydrous ≥97%
Tripotassium orthophosphate

CAS: 7778-53-2; EC No: 231-907-1; MDL No: MFCD00036295 Solid; Linear Formula: K3PO4; MW: 212.27 Melting Point: 1340° Density (g/mL): 2.564 Hygroscopic


  • Hygroscopic
  • Solid

Formula: K₃O₄P
MW: 212.27 g/mol
Melting Pt: 1340
Density: 2.564 g/mL
MDL Number: MFCD00036295
CAS Number: 7778-53-2
EINECS: 231-907-1
Merck Index: 12,07830

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR