2-Phenylbenzimidazole ≥97%
Supplier: BeanTown Chemical
Synonyms:
2-Phenyl-1H-benzimidazole, 2-Phenyl-1H-benzo[d]imidazole
128370-100G
128370-25G
128370-5G
BT128370-100GEA
189
USD
BT128370-100G
BT128370-25G
BT128370-5G
2-Phenylbenzimidazole ≥97%
2-Phenylbenzimidazole
CAS: 716-79-0; EC No: 211-939-2; MDL No: MFCD00005592; RTECS: DE0360000
Solid; Molecular Formula: C13H10N2; MW: 194.23
Melting Point: 293-296°
- RTECS: DE0360000
- Solid
Formula:
C₁₃H₁₀N₂ MW: 194.24 g/mol Melting Pt: 293…296 °C |
MDL Number:
MFCD00005592 CAS Number: 716-79-0 EINECS: 211-939-2 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...