L(+)-Serine ≥99%

Supplier: BeanTown Chemical

Synonyms: H-Ser-OH, (S)-2-Amino-3-hydroxypropanoic acid, Serine, (S)-(+)-Serine

128350-100G 128350-25G 128350-500G
BT128350-100GEA 93.04 USD
BT128350-100G BT128350-25G BT128350-500G
L(+)-Serine ≥99%
L(+)-Serine

CAS: 56-45-1; EC No: 200-274-3; MDL No: MFCD00064224; RTECS: VT8100000 Powder; Molecular Formula: C3H7NO3; MW: 105.09 Melting Point: 222° (decomposes) Optical Rotation: [α]20/D +13.0±0.5°, c = 5% in 5M HCl


  • RTECS: VT8100000
  • Optical rotation: [α]20/D +13.0±0.5°, c = 5% in 5M HCl
  • Powder

Formula: C₃H₇NO₃
MW: 105.09 g/mol
Boiling Pt: 259
Melting Pt: 222 °C (dec.)
MDL Number: MFCD00064224
CAS Number: 56-45-1
EINECS: 200-274-3
Merck Index: 14,00846

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR