4-Chloro-3-nitro-tert-butylbenzene

Supplier: BeanTown Chemical
Warning

Synonyms: 4-(tert-Butyl)-1-chloro-2-nitrobenzene

128345-1G 128345-5G
BT128345-1GEA 37 USD
BT128345-1G BT128345-5G
4-Chloro-3-nitro-tert-butylbenzene
4-Chloro-3-nitro-tert-butylbenzene

CAS: 58574-05-3; MDL No: MFCD07368382 Liquid; Linear Formula: ClC6H3(NO2)[C(CH3)3]; Molecular Formula: C10H12ClNO2; MW: 213.66


  • Liquid

Formula: C₁₀H₁₂ClNO₂
MW: 213.661 g/mol
MDL Number: MFCD07368382
CAS Number: 58574-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR