Succinimide ≥98%

Supplier: BeanTown Chemical

Synonyms: Pyrrolidine-2,5-dione

128325-1KG 128325-250G 128325-5KG
BT128325-1KGEA 124 USD
BT128325-1KG BT128325-250G BT128325-5KG
Succinimide ≥98%
Succinimide

CAS: 123-56-8; EC No: 204-635-6; MDL No: MFCD00005495; RTECS: WN2200000 Solid; Molecular Formula: C4H5NO2 ; MW: 99.09 Melting Point: 123-125°; Boiling Point: 285-290°; Flash point: 201°C (393°F) Density (g/mL): 1.41


  • RTECS: WN2200000
  • Solid

Formula: C₄H₅NO₂
MW: 99.09 g/mol
Boiling Pt: 285…290 °C
Melting Pt: 123…125 °C
Density: 1.41 g/mL
Flash Pt: 201 °C (393 °F)
MDL Number: MFCD00005495
CAS Number: 123-56-8
EINECS: 204-635-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR