Brij® 35

Supplier: BeanTown Chemical

Synonyms: Polyoxyethylene(23)lauryl ether, Tricosaethylene glycol mono-n-dodecyl ether, Brij® L23

128310-1KG 128310-250G 128310-5KG
BT128310-1KGEA 74 USD
BT128310-1KG BT128310-250G BT128310-5KG
Brij® 35
Brij® 35

CAS: 9002-92-0; EC No: 500-002-6; MDL No: MFCD00080891; RTECS: JR5990000 Liquid/Low Melting Solid


  • RTECS: JR5990000
  • Liquid/Low Melting Solid

Formula: C₅₈H₁₁₈O₂₄
MW: 1199.56 g/mol
MDL Number: MFCD00080891
CAS Number: 9002-92-0
EINECS: 500-002-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR