3-Octyn-1-ol ≥96%
Supplier: BeanTown Chemical
Synonyms:
Oct-3-yn-1-ol
128300-25G
128300-5G
BT128300-25GEA
171
USD
BT128300-25G
BT128300-5G
3-Octyn-1-ol ≥96%
3-Octyn-1-ol
CAS: 14916-80-4; EC No: 238-986-1; MDL No: MFCD00039552
Liquid; Linear Formula: CH3(CH2)3C≡C(CH2)2OH; Molecular Formula: C8H14O; MW: 126.20
Density (g/mL): 0.880; Refractive Index: 1.457
- Refractive index: 1.457
- Liquid
Formula:
C₈H₁₄O MW: 126.2 g/mol Density: 0.880 g/mL |
MDL Number:
MFCD00039552 CAS Number: 14916-80-4 EINECS: 238-986-1 |
Specification Test Results
Assay | 96% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...