Aluminium potassium sulfate dodecahydrate ≥98%

Supplier: BeanTown Chemical

128275-2.5KG 128275-500G
BT128275-2.5KGEA 263 USD
BT128275-2.5KG BT128275-500G
Aluminium potassium sulfate dodecahydrate ≥98%
Aluminium potassium sulphate dodecahydrate

CAS: 7784-24-9; EC No: 233-141-3; MDL No: MFCD00149143; RTECS: WS5650000 Solid; Linear Formula: KAl(SO4)2·12H2O ; MW: 474.39 Melting Point: 92° Density (g/mL): 1.757


  • RTECS: WS5650000
  • Solid

Formula: AlK(SO₄)₂·12H₂O
Melting Pt: 92 °C
Density: 1.757 g/mL
MDL Number: MFCD00149143
CAS Number: 7784-24-9
EINECS: 233-141-3

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR