(Methoxymethyl)triphenylphosphonium chloride ≥98%

Supplier: BeanTown Chemical

128255-100G 128255-25G 128255-500G
BT128255-100GEA 136 USD
BT128255-100G BT128255-25G BT128255-500G
(Methoxymethyl)triphenylphosphonium chloride ≥98%
(Methoxymethyl)triphenylphosphonium chloride

CAS: 4009-98-7; EC No: 223-664-5; MDL No: MFCD00011800 Crystalline/Powder; Linear Formula: CH3OCH2P(C6H5)3Cl; Molecular Formula: C20H20ClOP; MW: 342.80 Melting Point: 185-195° (decomposes) Hygroscopic


  • Hygroscopic
  • Crystalline/Powder

Formula: C₂₀H₂₀ClOP
MW: 342.8 g/mol
Melting Pt: 185…195 °C (dec.)
MDL Number: MFCD00011800
CAS Number: 4009-98-7
EINECS: 223-664-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR