Cerium tribromide ≥99.995%

Supplier: BeanTown Chemical

Synonyms: Cerous bromide, Cerium(III) bromide hydrate, Cerium (III) bromide

128150-10G 128150-50G
BT128150-10GEA 69 USD
BT128150-10G BT128150-50G
Cerium tribromide ≥99.995%
Cerium tribromide

CAS: 14457-87-5; EC No: 238-447-0; MDL No: MFCD00150206 Crystalline; Linear Formula: CeBr3·xH2O; MW: 379.83 (anhydrous)


  • Crystalline

Formula: CeBr₃
MW: 379.83 g/mol
Melting Pt: 732
MDL Number: MFCD00150206
CAS Number: 14457-87-5
EINECS: 238-447-0
Merck Index: 12,02039

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR