2-Ethylhexyl acrylate ≥98%

Supplier: BeanTown Chemical

128115-100ML 128115-500ML
BT128115-100MLEA 25 USD
BT128115-100ML BT128115-500ML
2-Ethylhexyl acrylate ≥98%
2-Ethylhexyl acrylate

CAS: 103-11-7; EC No: 203-080-7; MDL No: MFCD00009495; RTECS: AT0855000 Liquid; Molecular Formula: C11H20O2; MW: 184.28 Boiling Point: 215-219°; Flash point: 79°C (174°F) Density (g/mL): 0.885; Refractive Index: 1.436


  • RTECS: AT0855000
  • Refractive index: 1.436
  • Liquid

Formula: C₁₁H₂₀O₂
Boiling Pt: 215…219 °C
Density: 0.885 g/mL
Flash Pt: 79 °C (174 °F)
MDL Number: MFCD00009495
CAS Number: 103-11-7
EINECS: 203-080-7

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR