Platinum(II) acetylacetonate

Supplier: BeanTown Chemical

128095-1G 128095-5G
BT128095-1GEA 133 USD
BT128095-1G BT128095-5G
Platinum(II) acetylacetonate
Platinum(II) acetylacetonate

CAS: 15170-57-7; EC No: 239-223-5; MDL No: MFCD00000028 Needles; Linear Formula: Pt(C5H7O2)2; MW: 393.30 Melting Point: 249-252°


  • Needles

Formula: C₁₀H₁₄O₄Pt
Melting Pt: 249…252 °C
MDL Number: MFCD00000028
CAS Number: 15170-57-7
EINECS: 239-223-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR