5-Aminoisophthalic acid ≥95%
Supplier: BeanTown Chemical
128085-250G
128085-50G
BT128085-250GEA
213
USD
BT128085-250G
BT128085-50G
5-Aminoisophthalic acid ≥95%
5-Aminoisophthalic acid
CAS: 99-31-0; EC No: 202-748-5; MDL No: MFCD00002522
Powder; Molecular Formula: C8H7NO4; MW: 181.15
Melting Point:
- Powder
Formula:
C₈H₇NO₄ Melting Pt: >300 °C |
MDL Number:
MFCD00002522 CAS Number: 99-31-0 EINECS: 202-748-5 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...