D-tert-Leucine (H-D-Tle-OH) ≥99%

Supplier: BeanTown Chemical

Synonyms: H-D-Tle-OH

128030-100MG 128030-2G 128030-500MG
BT128030-100MGEA 58 USD
BT128030-100MG BT128030-2G BT128030-500MG
D-tert-Leucine (H-D-Tle-OH) ≥99%
D-tert-Leucine (H-D-Tle-OH)

CAS: 26782-71-8; MDL No: MFCD00004265 Crystalline; Molecular Formula: C6H13NO2; MW: 131.17 Optical Rotation: [α]25/D +9.5°, c = 3% in water


  • Optical rotation: [α]25/D +9.5°, c = 3% in water
  • Crystalline

Formula: C₆H₁₃NO₂
MW: 131.2 g/mol
MDL Number: MFCD00004265
CAS Number: 26782-71-8

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR