N-(p-Toluenesulfonyl)glycine ≥97%

Supplier: BeanTown Chemical

Synonyms: N-Tosylglycine, N-p-Tosylglycine, Tos-Gly-OH

128010-25G 128010-5G
BT128010-25GEA 142 USD
BT128010-25G BT128010-5G
N-(p-Toluenesulfonyl)glycine ≥97%
N-(p-Toluenesulfonyl)glycine

CAS: 1080-44-0; MDL No: MFCD00045898 Solid; Linear Formula: CH3C6H4NHCH2COOH; Molecular Formula: C9H11NO4S ; MW: 229.25 Melting Point: 147-149°


  • Solid

Formula: C₉H₁₁NO₄S
MW: 229.25 g/mol
Melting Pt: 147…149 °C
MDL Number: MFCD00045898
CAS Number: 1080-44-0

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR