(R)-(-)-Thiazolidine-4-carboxylic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
H-Thz-OH, L(-)-Thiazolidine-4-carboxylic acid
127970-10G
127970-250G
127970-50G
BT127970-10GEA
23
USD
BT127970-10G
BT127970-250G
BT127970-50G
(R)-(-)-Thiazolidine-4-carboxylic acid ≥98%
(R)-(-)-Thiazolidine-4-carboxylic acid
CAS: 34592-47-7; EC No: 207-146-6; MDL No: MFCD00005212; RTECS: XJ5425500
Crystalline; Molecular Formula: C4H7NO2S ; MW: 133.17
Melting Point: 190-200° (decomposes)
Optical Rotation: [α]20/D -141°, c = 1.3 in water
- RTECS: XJ5425500
- Optical rotation: [α]20/D -141°, c = 1.3 in water
- Crystalline
Formula:
C₄H₇NO₂S Melting Pt: 190…200 °C (dec.) |
MDL Number:
MFCD00005212 CAS Number: 34592-47-7 EINECS: 207-146-6 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...