L(+)-Asparagine monohydrate ≥98%

Supplier: BeanTown Chemical

Synonyms: L-Asparagine monohydrate, H-Asn-OH·H₂O, (S)-(+)-Asparagine monohydrate, (S)-2,4-Diamino-4-oxobutanoic acid hydrate, H-Asn-OH.H2O

127965-100G 127965-25G 127965-500G
BT127965-100GEA 50.82 USD
BT127965-100G BT127965-25G BT127965-500G
L(+)-Asparagine monohydrate ≥98%
L(+)-Asparagine monohydrate

CAS: 5794-13-8; EC No: 200-735-9; MDL No: MFCD00151038 Powder; Molecular Formula: C27H18Cl3N3O; MW: 150.13 Melting Point: 233-235°


  • Powder

Formula: C₄H₈N₂O₃·1H₂O
MW: 150.13 g/mol
Melting Pt: 233…235 °C
MDL Number: MFCD00151038
CAS Number: 5794-13-8
EINECS: 200-735-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR