Alginic acid

Supplier: BeanTown Chemical

127870-100G 127870-2.5KG 127870-500G
BT127870-100GEA 47.07 USD
BT127870-100G BT127870-2.5KG BT127870-500G
Alginic acid
Alginic acid

CAS: 9005-32-7; EC No: 232-680-1; MDL No: MFCD00081309; RTECS: AZ5775000 Solid; Molecular Formula: (C6H8O6)n Hygroscopic


  • RTECS: AZ5775000
  • Hygroscopic
  • Solid

Formula: (C₆H₈O₆)n
MW: 176.13 g/mol
MDL Number: MFCD00081309
CAS Number: 9005-32-7
EINECS: 232-680-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR