1-(Phenylsulfonyl)pyrrole ≥98%

Supplier: BeanTown Chemical

127860-1G 127860-25G 127860-5G
BT127860-1GEA 35 USD
BT127860-1G BT127860-25G BT127860-5G
1-(Phenylsulfonyl)pyrrole ≥98%
1-(Phenylsulfonyl)pyrrole

CAS: 16851-82-4; MDL No: MFCD00067739 Solid; Molecular Formula: C10H9NO2S; MW: 207.25 Melting Point: 88-91°


  • Solid

Formula: C₁₀H₉NO₂S
MW: 207.247 g/mol
Melting Pt: 88…91 °C
MDL Number: MFCD00067739
CAS Number: 16851-82-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR