2,3,4-Trimethoxybenzaldehyde ≥98%

Supplier: BeanTown Chemical

127845-10G 127845-250G 127845-50G
BT127845-10GEA 31 USD
BT127845-10G BT127845-250G BT127845-50G
2,3,4-Trimethoxybenzaldehyde ≥98%
2,3,4-Trimethoxybenzaldehyde

CAS: 2103-57-3; EC No: 218-271-0; MDL No: MFCD00003310 Crystalline/Powder; Molecular Formula: C10H12O4; MW: 196.20 Melting Point: 38-40°; Boiling Point: 168-170°/12 mmHg; Flash point: 110°C (230°F) Refractive Index: 1.555 Air Sensitive


  • Refractive index: 1.555
  • Air Sensitive
  • Crystalline/Powder

Formula: C₁₀H₁₂O₄
MW: 196.2 g/mol
Boiling Pt: 168…170 °C/12 mmHg
Melting Pt: 38…40 °C
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00003310
CAS Number: 2103-57-3
EINECS: 218-271-0

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR