Fluorescein disodium salt

Supplier: BeanTown Chemical

Synonyms: 3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, disodium salt, Acid Yellow 73, Yellow No. 8, Fluorescein sodium salt, 2-(6-hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid disodium salt, Disodium 2-(3-oxo-6-oxidoxanthen-9-yl)benzoate

127830-100G 127830-2.5KG 127830-500G
BT127830-100GEA 30.48 USD
BT127830-100G BT127830-2.5KG BT127830-500G
Fluorescein disodium salt
Fluorescein disodium salt

CAS: 518-47-8; EC No: 208-253-0; MDL No: MFCD00167039; RTECS: LM5425000 Powder; Molecular Formula: C20H10Na2O5; MW: 376.28


  • RTECS: LM5425000
  • Powder

Formula: C₂₀H₁₀Na₂O₅
MW: 376.28 g/mol
MDL Number: MFCD00167039
CAS Number: 518-47-8
EINECS: 208-253-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=949f2473a2f0c6f8d00fd80ca2f80b730bc8076a&vid=fe174603ab8c1727364c1e09d665a1ad5a6d9106&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR