Methyl-3-amino-5,6-dichloropyrazine-2-carboxylate ≥97%

Supplier: BeanTown Chemical

Synonyms: Methyl 3-amino-5,6-dichloropyrazine-2-carboxylate

127825-1G 127825-5G
BT127825-1GEA 79 USD
BT127825-1G BT127825-5G
Methyl-3-amino-5,6-dichloropyrazine-2-carboxylate ≥97%
Methyl 3-amino-5,6-dichloropyrazine-2-carboxylate

CAS: 1458-18-0; EC No: 215-947-7; MDL No: MFCD00010431 Powder; Molecular Formula: C6H5Cl2N3O2; MW: 222.03 Melting Point: 230-233°


  • Powder

Formula: C₆H₅Cl₂N₃O₂
MW: 222.03 g/mol
Melting Pt: 230…233 °C
MDL Number: MFCD00010431
CAS Number: 1458-18-0
EINECS: 215-947-7

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR