Ammonium 1-pyrrolidinecarbodithioate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Pyrrolidinedithiocarbamate ammonium salt, 1-Pyrrolidinecarbodithioic acid, ammonium salt, Ammonium pyrrolidine-1-carbodithioate, Pyrrolidine-1-Dithiocarboxylic Acid Ammonium Salt, APCD
127820-100G
127820-25G
BT127820-100GEA
167
USD
BT127820-100G
BT127820-25G
Ammonium 1-pyrrolidinecarbodithioate ≥98%
Ammonium 1-pyrrolidinecarbodithioate
CAS: 5108-96-3; EC No: 225-834-4; MDL No: MFCD00012720
Crystalline/Solid; Molecular Formula: C5H12N2S2; MW: 164.29
Melting Point: 153-155°
Hygroscopic
- Hygroscopic
- Crystalline/Solid
Formula:
C₅H₁₂N₂S₂ MW: 146.3 · 18.0 g/mol Melting Pt: 153…155 °C |
MDL Number:
MFCD00012720 CAS Number: 5108-96-3 EINECS: 225-834-4 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...