Lovastatin ≥97%
Supplier: BeanTown Chemical
Synonyms:
6alpha-Methylcompactin, Monacolin K, Mevinolin, (S)-(1S,3R,7S,8S,8aR)-8-(2-((2R,4R)-4-Hydroxy-6-oxotetrahydro-2H-pyran-2-yl)ethyl)-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl 2-methylbutanoate
127790-25G
127790-5G
BT127790-25GEA
262
USD
BT127790-25G
BT127790-5G
Lovastatin ≥97%
Lovastatin
CAS: 75330-75-5; MDL No: MFCD00072164; RTECS: EK7907000
Crystalline/Powder; Molecular Formula: C24H36O5; MW: 404.54
- RTECS: EK7907000
- Crystalline/Powder
Formula:
C₂₄H₃₆O₅ MW: 404.55 g/mol |
MDL Number:
MFCD00072164 CAS Number: 75330-75-5 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...