Salicylaldehyde azine ≥97%

Supplier: BeanTown Chemical

Synonyms: Bis(2-hydroxybenzaldehyde) hydrazone, N,N'-Bis(salicylidene)hydrazine, Salazine, Salicylaldazine, 2,2'-(Hydrazine-1,2-diylidenebis(methanylylidene))diphenol

127680-10G 127680-50G
BT127680-10GEA 100.2 USD
BT127680-10G BT127680-50G
Salicylaldehyde azine ≥97%
Salicylaldehyde azine

CAS: 959-36-4; EC No: 213-500-0; MDL No: MFCD00043496 Crystalline/Powder; Linear Formula: HOC6H4CH=NN=CHC6H4OH; Molecular Formula: C14H12N2O2; MW: 240.26 Melting Point: 216-222°


  • Crystalline/Powder

Formula: C₁₄H₁₂N₂O₂
MW: 240.26 g/mol
Melting Pt: 216…222 °C
MDL Number: MFCD00043496
CAS Number: 959-36-4
EINECS: 213-500-0

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR