Phenethyl bromide ≥98%
Supplier: BeanTown Chemical
Synonyms:
(2-Bromoethyl)benzene, 2-Phenylethyl bromide
127665-100G
127665-500G
BT127665-100GEA
36.49
USD
BT127665-100G
BT127665-500G
Phenethyl bromide ≥98%
Phenethyl bromide
CAS: 103-63-9; EC No: 203-130-8; MDL No: MFCD00000240; RTECS: CY9032000
Liquid; Linear Formula: C6H5(CH2)2Br; MW: 185.06
Boiling Point: 220-221°; Flash point: 96°C (205°F)
Density (g/mL): 1.355; Refractive Index: 1.556
- RTECS: CY9032000
- Refractive index: 1.556
- Liquid
Formula:
BrCH₂CH₂C₆H₅ MW: 185.06 g/mol Boiling Pt: 220…221 °C Density: 1.355 g/mL Flash Pt: 96 °C (205 °F) |
MDL Number:
MFCD00000240 CAS Number: 103-63-9 EINECS: 203-130-8 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...