Methyl red ACS
Supplier: BeanTown Chemical
127630-100G
127630-25G
BT127630-100GEA
81.85
USD
BT127630-100G
BT127630-25G
Methyl red ACS
Methyl red
CAS: 493-52-7; EC No: 207-776-1; MDL No: MFCD00002425; RTECS: DG8960000
Crystalline; Linear Formula: (CH3)2NC6H4N=NC6H4COOH; Molecular Formula: C15H15N3O2 ; MW: 269.30
Melting Point: 179-182°
- RTECS: DG8960000
- Crystalline
Formula:
C₁₅H₁₅N₃O₂ Melting Pt: 179…182 °C |
MDL Number:
MFCD00002425 CAS Number: 493-52-7 EINECS: 207-776-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...