trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate 97.5-100.5% ACS

Supplier: BeanTown Chemical

Synonyms: Titriplex® IV

127600-100G 127600-25G 127600-5G
BT127600-100GEA 248 USD
BT127600-100G BT127600-25G BT127600-5G
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate 97.5-100.5% ACS
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate

CAS: 125572-95-4; EC No: 236-308-9; MDL No: MFCD00066429 Solid; Molecular Formula: C14H22N2O8·H2O ; MW: 364.35 Melting Point: 213-216°


  • Solid

Formula: C₁₄H₂₂N₂O₈·1H₂O
MW: 364.35 g/mol
Melting Pt: 213…216 °C
MDL Number: MFCD00066429
CAS Number: 125572-95-4
EINECS: 236-308-9

Order Now

Specification Test Results

Assay 97.5-100.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR