Dysprosium(III) oxide ≥99.9% (trace rare earth metals basis)
Supplier: BeanTown Chemical
Synonyms:
Didysprosium trioxide, Dysprosium oxide
127570-10G
127570-250G
127570-50G
BT127570-10GEA
49
USD
BT127570-10G
BT127570-250G
BT127570-50G
Dysprosium(III) oxide ≥99.9% (trace rare earth metals basis)
Dysprosium(III) oxide
CAS: 1308-87-8; EC No: 215-164-0; MDL No: MFCD00010986
Powder; Molecular Formula: Dy2O3; MW: 373.00
Density (g/mL): 7.81
Hygroscopic
- Hygroscopic
- Powder
Formula:
Dy₂O₃ MW: 373 g/mol Density: 7.81 g/mL |
MDL Number:
MFCD00010986 CAS Number: 1308-87-8 EINECS: 215-164-0 |
Specification Test Results
Assay | 99.9% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...