Cerium zirconium oxide ≥99.5% (trace metals basis)

Supplier: BeanTown Chemical

127550-100G 127550-500G
BT127550-100GEA 89 USD
BT127550-100G BT127550-500G
Cerium zirconium oxide ≥99.5% (trace metals basis)
Cerium zirconium oxide

CAS: 53169-24-7; EC No: 258-407-6; MDL No: MFCD00168089 -325 Mesh Powder; Molecular Formula: CeO2·ZrO2 ; MW: 295.34


  • -325 Mesh Powder

Formula: CeO₂·ZrO₂
MW: 295.32 g/mol
MDL Number: MFCD00168089
CAS Number: 53169-24-7
EINECS: 258-407-6

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR