N-Carbethoxy-4-piperidone ≥99%
Supplier: BeanTown Chemical
Synonyms:
4-Piperidone-N-carboxylic acid ethyl ester, N-Ethoxycarbonyl-4-piperidone, Ethyl4-oxo-1-piperidinecarboxylate
127515-10G
127515-250G
127515-50G
BT127515-10GEA
38
USD
BT127515-10G
BT127515-250G
BT127515-50G
N-Carbethoxy-4-piperidone ≥99%
N-Carbethoxy-4-piperidone
CAS: 29976-53-2; EC No: 249-984-5; MDL No: MFCD00006188
Liquid; Molecular Formula: C8H13NO3; MW: 171.20
Boiling Point: 95-98°/1 mmHg; Flash point: 87°C (189°F)
Density (g/mL): 1.135; Refractive Index: 1.475
- Refractive index: 1.475
- Liquid
Formula:
C₈H₁₃NO₃ MW: 171.19 g/mol Boiling Pt: 95…98 °C/1 mmHg Density: 1.135 g/mL Flash Pt: 87 °C (189 °F) |
MDL Number:
MFCD00006188 CAS Number: 29976-53-2 EINECS: 249-984-5 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...