Chromotropic acid disodium salt dihydrate ACS

Supplier: BeanTown Chemical

Synonyms: 4,5-Dihydroxynaphthalene-2,7-disulphonic acid disodium salt dihydrate, Disodium 4,5-dihydroxynaphthalene-2,7-disulphonate dihydrate, 4,5-Dihydroxynaphthalene-2,7-disulphonic acid, disodium salt dihydrate, 4,5-Dihydroxy-2,7-naphthalenedisulphonic acid disodium salt dihydrate

127450-100G 127450-25G 127450-5G
BT127450-100GEA 207 USD
BT127450-100G BT127450-25G BT127450-5G
Chromotropic acid disodium salt dihydrate ACS
Chromotropic acid disodium salt dihydrate

CAS: 5808-22-0; EC No: 204-972-9; MDL No: MFCD00150612 Powder; Molecular Formula: C10H6Na2O8S2·2H2O; MW: 400.29 Melting Point: 300°


  • Powder

Formula: (HO)₂C₁₀H₄(SO₃Na)₂·2H₂O
MW: 400.29 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00150612
CAS Number: 5808-22-0
EINECS: 204-972-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR