Trichloroacetamide ≥99%

Supplier: BeanTown Chemical

Synonyms: 2,2,2-Trichloroacetamide

127440-250G 127440-50G
BT127440-250GEA 135 USD
BT127440-250G BT127440-50G
Trichloroacetamide ≥99%
Trichloroacetamide

CAS: 594-65-0; EC No: 209-849-3; MDL No: MFCD00008009; RTECS: AC9275000 Crystalline; Linear Formula: Cl3CCONH2 ; MW: 162.40 Melting Point: 139-141°; Boiling Point: 238-240°


  • RTECS: AC9275000
  • Crystalline

Formula: C₂H₂Cl₃NO
MW: 162.4 g/mol
Boiling Pt: 238…240 °C
Melting Pt: 139…141 °C
MDL Number: MFCD00008009
CAS Number: 594-65-0
EINECS: 209-849-3

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR